Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-4-methoxyindole-2-carboxylate 97%

Supplier: AMBEED, INC
Warning

A126457-100MG A126457-1G A126457-250MG A126457-5G
76718-734EA 11 USD InStock
76718-734 76718-736 76718-738 76718-740
Methyl-4-methoxyindole-2-carboxylate 97%
Methyl-4-methoxyindole-2-carboxylate
Precaución: for Research Use Only


Formula: C₁₁H₁₁NO₃
MW: 205.21 g/mol
MDL Number: MFCD00134300
CAS Number: 111258-23-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty