Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dimethyl phenylphosphonate 98%

Supplier: AMBEED, INC
Warning

A126579-10G A126579-1G A126579-25G A126579-5G
76718-756EA 24 USD InStock
76718-756 76718-758 76718-760 76718-762
Dimethyl phenylphosphonate 98%
Dimethyl phenylphosphonate
Precaución: for Research Use Only


Formula: C₈H₁₁O₃P
MDL Number: MFCD00059292
CAS Number: 2240-41-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty