Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Quipazine dimaleate salt 95%

Supplier: AMBEED, INC
Warning

A127024-100MG A127024-250MG
76718-796EA 31 USD InStock
76718-796 77592-464
Quipazine dimaleate salt 95%
Quipazine dimaleate salt
Precaución: for Research Use Only


Formula: C₁₃H₁₅N₃
MW: 213.284 g/mol
MDL Number: MFCD00242605
CAS Number: 4774-24-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty