Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(2R)-2-Amino-3-methylsulfinyl-propanoic acid 97%

Supplier: AMBEED, INC
Warning

A131282-100MG A131282-250MG A131282-5G A131282-1G
76718-950EA 8 USD InStock
76718-950 76718-952 76718-954 77666-884
(2R)-2-Amino-3-methylsulfinyl-propanoic acid 97%
(2R)-2-Amino-3-methylsulfinyl-propanoic acid
Precaución: for Research Use Only


Formula: C₄H₉NO₃S
MDL Number: MFCD00152132
CAS Number: 6853-87-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty