Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

A77 1726 99+%

Supplier: AMBEED, INC
Warning

A132118-100MG A132118-1G A132118-1MG A132118-25MG
76718-982EA 23 USD InStock
76718-982 76718-984 76718-986 76718-988
A77 1726 99+%
A77 1726
Precaución: for Research Use Only


Formula: C₁₂H₉F₃N₂O₂
MDL Number: MFCD00910058
CAS Number: 108605-62-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty