Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4-Dihydroxybenzophenone 98%

Supplier: AMBEED, INC
Warning

A133469-100G A133469-25G A133469-500G
76719-036EA 28 USD InStock
76719-036 76719-038 76719-040
2,4-Dihydroxybenzophenone 98%
2,4-Dihydroxybenzophenone
Precaución: for Research Use Only


Formula: C₁₃H₁₀O₃
MDL Number: MFCD00002277
CAS Number: 131-56-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty