Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Penciclovir 98%

Supplier: AMBEED, INC
Warning

A135007-100MG A135007-50MG A135007-5G A135007-100G
76719-114EA 7 USD InStock
76719-114 76719-116 76719-118 77666-950
Penciclovir 98%
Penciclovir
Precaución: for Research Use Only


Formula: C₁₀H₁₅N₅O₃
MW: 253.26 g/mol
MDL Number: MFCD15142866
CAS Number: 39809-25-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty