Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Bromo-3,5-dimethyladamantane 98%

Supplier: AMBEED, INC
Warning

A135676-100G A135676-10G A135676-1G A135676-25G A135676-5G
76719-152EA 39 USD InStock
76719-152 76719-154 76719-156 76719-158 76719-160
1-Bromo-3,5-dimethyladamantane 98%
1-Bromo-3,5-dimethyladamantane
Precaución: for Research Use Only


Formula: C₁₂H₁₉Br
MDL Number: MFCD00077197
CAS Number: 941-37-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty