Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

SL327 98+%

Supplier: AMBEED, INC
Warning

A139600-10MG A139600-1MG A139600-5MG
76719-306EA 35 USD InStock
76719-306 76719-308 76719-310
SL327 98+%
SL327
Precaución: for Research Use Only


Formula: C₁₆H₁₂F₃N₃S
MDL Number: MFCD06411432
CAS Number: 305350-87-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty