Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,1-Dimethylpiperidin-1-ium-4-yloctadecyl phosphate 98%

Supplier: AMBEED, INC
Warning

A140384-100MG A140384-10MG A140384-1MG A140384-50MG A140384-5MG
76719-334EA 112 USD InStock
76719-334 76719-336 76719-338 76719-340 76719-342
1,1-Dimethylpiperidin-1-ium-4-yloctadecyl phosphate 98%
1,1-Dimethylpiperidin-1-ium-4-yloctadecyl phosphate
Precaución: for Research Use Only


Formula: C₂₅H₅₂NO₄P
MDL Number: MFCD00927554
CAS Number: 157716-52-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty