Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4',5'-Dichloro-2'-nitroacetanilide 97%

Supplier: AMBEED, INC
Warning

A141097-100G A141097-1G A141097-25G A141097-5G
76719-356EA 264 USD InStock
76719-356 76719-358 76719-360 76719-362
4',5'-Dichloro-2'-nitroacetanilide 97%
4',5'-Dichloro-2'-nitroacetanilide
Precaución: for Research Use Only


Formula: C₈H₆Cl₂N₂O₃
MDL Number: MFCD00024301
CAS Number: 5462-30-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty