Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Olopatadine hydrochloride 98%

Supplier: AMBEED, INC
Danger

A143866-10MG A143866-250MG A143866-25G A143866-50MG A143866-1MG
76719-408EA 6 USD InStock
76719-408 76719-410 76719-412 76719-414 77667-076
Olopatadine hydrochloride 98%
Olopatadine hydrochloride
Precaución: for Research Use Only


Formula: C₂₁H₂₃NO₃
MW: 373.88 g/mol
MDL Number: MFCD00875716
CAS Number: 140462-76-6
UN: 2811
ADR: 6.1 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty