Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-2-chloro-5-fluoronicotinate 97%

Supplier: AMBEED, INC
Warning

A144221-100MG A144221-10G A144221-1G A144221-250MG A144221-5G
76719-444EA 5 USD InStock
76719-444 76719-446 76719-448 76719-450 76719-452
Methyl-2-chloro-5-fluoronicotinate 97%
Methyl-2-chloro-5-fluoronicotinate
Precaución: for Research Use Only


Formula: C₇H₅ClFNO₂
MDL Number: MFCD09763661
CAS Number: 847729-27-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty