Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-(2-Aminoethylamino)-1-naphthalenesulfonic acid 95%

Supplier: AMBEED, INC
Warning

A144696-100MG A144696-1G A144696-250MG
76719-458EA 76 USD InStock
76719-458 76719-460 76719-462
5-(2-Aminoethylamino)-1-naphthalenesulfonic acid 95%
5-(2-Aminoethylamino)-1-naphthalenesulfonic acid
Precaución: for Research Use Only


Formula: C₁₂H₁₄N₂O₃S
MDL Number: MFCD00066932
CAS Number: 50402-56-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty