Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-(4-Bromo-2,6-dichlorophenyl)acetamide 98%

Supplier: AMBEED, INC
Warning

A147268-100MG A147268-1G A147268-250MG A147268-5G
76719-574EA 6 USD InStock
76719-574 76719-576 76719-578 76719-580
N-(4-Bromo-2,6-dichlorophenyl)acetamide 98%
N-(4-Bromo-2,6-dichlorophenyl)acetamide
Precaución: for Research Use Only


Formula: C₈H₆BrCl₂NO
MDL Number: MFCD08703152
CAS Number: 13953-09-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty