Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N(α)-Cbz-DL-tryptophan 98%

Supplier: AMBEED, INC
Warning

A154987-10G A154987-5G A154987-100G
76719-906EA 9 USD InStock
76719-906 76719-908 77667-194
N(α)-Cbz-DL-tryptophan 98%
N(α)-Cbz-DL-tryptophan
Precaución: for Research Use Only


Formula: C₁₉H₁₈N₂O₄
MW: 338.4 g/mol
MDL Number: MFCD00069707
CAS Number: 13058-16-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty