Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-5-formylthiophene-2-carboxylate 98%

Supplier: AMBEED, INC
Warning

A156236-10G A156236-1G A156236-250MG A156236-25G A156236-5G
76719-952EA 217 USD InStock
76719-952 76719-954 76719-956 76719-958 76719-960
Methyl-5-formylthiophene-2-carboxylate 98%
Methyl-5-formylthiophene-2-carboxylate
Precaución: for Research Use Only


Formula: C₇H₆O₃S
MDL Number: MFCD01859942
CAS Number: 67808-64-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty