Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Nebularine 98%

Supplier: AMBEED, INC
Warning

A157458-100MG A157458-250MG A157458-25MG A157458-50MG
76719-970EA 54 USD InStock
76719-970 76719-972 76719-974 77667-220
Nebularine 98%
Nebularine
Precaución: for Research Use Only


Formula: C₁₀H₁₂N₄O₄
MDL Number: MFCD00022819
CAS Number: 550-33-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty