Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,4-Difluorobenzophenone 98%

Supplier: AMBEED, INC
Warning

A158473-100G A158473-10G A158473-1G A158473-25G A158473-5G
76720-018EA 122 USD InStock
76720-018 76720-020 76720-022 76720-024 76720-026
3,4-Difluorobenzophenone 98%
3,4-Difluorobenzophenone
Precaución: for Research Use Only


Formula: C₁₃H₈F₂O
MDL Number: MFCD00009892
CAS Number: 85118-07-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty