Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-5-amino-2,4-difluorobenzoate 98%

Supplier: AMBEED, INC
Warning

A160280-100G A160280-10G A160280-1G A160280-25G A160280-5G
76720-104EA 258 USD InStock
76720-104 76720-106 76720-108 76720-110 76720-112
Methyl-5-amino-2,4-difluorobenzoate 98%
Methyl-5-amino-2,4-difluorobenzoate
Precaución: for Research Use Only


Formula: C₈H₇F₂NO₂
MDL Number: MFCD09396710
CAS Number: 125568-73-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty