Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Norfloxacin 98+%

Supplier: AMBEED, INC
Warning

A165613-100G A165613-10G A165613-25G A165613-500G A165613-50MG A165613-5G
76720-284EA 58 USD InStock
76720-284 76720-286 76720-288 76720-290 76720-292 76720-294
Norfloxacin 98+%
Norfloxacin
Precaución: for Research Use Only


Formula: C₁₆H₁₈FN₃O₃
MDL Number: MFCD00079532
CAS Number: 70458-96-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty