Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-(2-Aminoethyl)glycine 97%

Supplier: AMBEED, INC
Warning

A175646-100MG A175646-250MG A175646-5G
76720-652EA 26 USD InStock
76720-652 76720-654 76720-656
N-(2-Aminoethyl)glycine 97%
N-(2-Aminoethyl)glycine
Precaución: for Research Use Only


Formula: C₄H₁₀N₂O₂
MW: 118.14 g/mol
MDL Number: MFCD00144824
CAS Number: 24123-14-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty