Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(Chloromethyl)-2,3,4,5,6-pentafluorobenzene ≥98%

Supplier: AMBEED, INC
Danger

A179554-1G A179554-5G
76720-822EA 79 USD InStock
76720-822 76720-824
1-(Chloromethyl)-2,3,4,5,6-pentafluorobenzene ≥98%
1-(Chloromethyl)-2,3,4,5,6-pentafluorobenzene
Precaución: for Research Use Only


Formula: C₇H₂ClF₅
MW: 216.54 g/mol
MDL Number: MFCD00040864
CAS Number: 653-35-0
UN: 2922
ADR: 8,6.1 Ⅱ
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty