Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-4-[amino(hydroxyimino)methyl]benzenecarboxylate 97%

Supplier: AMBEED, INC
Warning

A181383-1G A181383-250MG A181383-25G
76720-890EA 25 USD InStock
76720-890 76720-892 77667-504
Methyl-4-[amino(hydroxyimino)methyl]benzenecarboxylate 97%
Methyl-4-[amino(hydroxyimino)methyl]benzenecarboxylate
Precaución: for Research Use Only


Formula: C₉H₁₀N₂O₃
MDL Number: MFCD08689689
CAS Number: 65695-05-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty