Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(Phenylsulfonyl)aniline 98+%

Supplier: AMBEED, INC
Warning

A181682-100G A181682-25G A181682-500G A181682-5G
76720-894EA 48 USD InStock
76720-894 76720-896 76720-898 76720-900
2-(Phenylsulfonyl)aniline 98+%
2-(Phenylsulphonyl)aniline
Precaución: for Research Use Only


Formula: C₁₂H₁₁NO₂S
MDL Number: MFCD00007706
CAS Number: 4273-98-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty