Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl decanoate 98%

Supplier: AMBEED, INC
Danger

A181864-100G A181864-500G
76720-912EA 9 USD InStock
76720-912 76720-914
Methyl decanoate 98%
Methyl decanoate
Precaución: for Research Use Only


Formula: C₁₁H₂₂O₂
MDL Number: MFCD00009580
CAS Number: 110-42-9
UN: 3082
ADR: 9,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty