Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl toluene-4-sulphonate 98%

Supplier: AMBEED, INC
Danger

A182547-1000G A182547-100G A182547-25G A182547-500G
76720-940EA 44 USD InStock
76720-940 76720-942 76720-944 76720-946
Methyl toluene-4-sulphonate 98%
Methyl toluene-4-sulphonate
Precaución: for Research Use Only


Formula: C₈H₁₀O₃S
MW: 186.23 g/mol
Boiling Pt: 292
Flash Pt: 130
MDL Number: MFCD00008417
CAS Number: 80-48-8
UN: 3265
ADR: 8 Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty