Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(Diethylamino)-2-methoxybenzaldehyde 95%

Supplier: AMBEED, INC
Warning

A182654-10G A182654-1G A182654-250MG A182654-25G
76720-948EA 96 USD InStock
76720-948 76720-950 76720-952 76720-954
4-(Diethylamino)-2-methoxybenzaldehyde 95%
4-(Diethylamino)-2-methoxybenzaldehyde
Precaución: for Research Use Only


Formula: C₁₂H₁₇NO₂
MDL Number: MFCD02277405
CAS Number: 55586-68-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty