Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

S-Methyl-2-furancarbothioate 95%

Supplier: AMBEED, INC
Warning

A183442-100G A183442-25G A183442-500G A183442-5G
76720-968EA 92 USD InStock
76720-968 76720-970 76720-972 76720-974
S-Methyl-2-furancarbothioate 95%
S-Methyl-2-furancarbothioate
Precaución: for Research Use Only


Formula: C₆H₆O₂S
MDL Number: MFCD00040266
CAS Number: 13679-61-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty