Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Plumbagin 97%

Supplier: AMBEED, INC
Warning

A187738-100MG A187738-10MG A187738-1MG
76721-064EA 76 USD InStock
76721-064 76721-066 76721-068
Plumbagin 97%
Plumbagin
Precaución: for Research Use Only


Formula: C₁₁H₈O₃
MDL Number: MFCD00001682
CAS Number: 481-42-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty