Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Nicotinamide-N-oxide 98%

Supplier: AMBEED, INC
Warning

A190426-100G A190426-100MG A190426-25G A190426-500G A190426-1MG A190426-5G
76721-120EA 120 USD InStock
76721-120 76721-122 76721-124 76721-126 77667-590 77667-592
Nicotinamide-N-oxide 98%
Nicotinamide-N-oxide
Precaución: for Research Use Only


Formula: C₆H₆N₂O₂
MDL Number: MFCD00006202
CAS Number: 1986-81-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty