Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

PIPES disodium salt 98%

Supplier: AMBEED, INC
Warning

A192121-1000G A192121-100G A192121-500G
76721-144EA 80 USD InStock
76721-144 76721-146 76721-148 76721-150
PIPES disodium salt 98%
PIPES disodium salt
Precaución: for Research Use Only


Formula: C₈H₁₆N₂Na₂O₆S₂
MW: 346.34 g/mol
MDL Number: MFCD00064349
CAS Number: 76836-02-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty