Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Phthalylsulfacetamide 98%

Supplier: AMBEED, INC
Warning

A194488-100MG A194488-1MG A194488-250MG
76721-238EA 11 USD InStock
76721-238 76721-240 76721-242
Phthalylsulfacetamide 98%
Phthalylsulfacetamide
Precaución: for Research Use Only


Formula: C₁₆H₁₄N₂O₆S
MDL Number: MFCD00020275
CAS Number: 131-69-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty