Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 4-Benzoylbutyrate 97%

Supplier: AMBEED, INC
Warning

A196542-1G A196542-250MG A196542-25G A196542-5G
76721-306EA 31 USD InStock
76721-306 76721-308 76721-310 76721-312
Methyl 4-Benzoylbutyrate 97%
Methyl 4-Benzoylbutyrate
Precaución: for Research Use Only


Formula: C₁₂H₁₄O₃
MDL Number: MFCD00043757
CAS Number: 1501-04-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty