Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dibenzothiophene-2-boronic acid 97%

Supplier: AMBEED, INC
Warning

A198359-10G A198359-1G A198359-250MG A198359-5G
76721-356EA 27 USD InStock
76721-356 76721-358 76721-360 76721-362
Dibenzothiophene-2-boronic acid 97%
Dibenzothiophene-2-boronic acid
Precaución: for Research Use Only


Formula: C₁₂H₉BO₂S
MDL Number: MFCD01318982
CAS Number: 668983-97-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty