Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Neostigmine bromide 98%

Supplier: AMBEED, INC
Danger

A198984-100MG A198984-25MG
76721-384EA 8 USD InStock
76721-384 76721-386
Neostigmine bromide 98%
Neostigmine bromide
Precaución: for Research Use Only


Formula: C₁₂H₁₉BrN₂O₂
MDL Number: MFCD00011795
CAS Number: 114-80-7
UN: 2811
ADR: 6.1,Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty