Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-3-methoxy-2-naphthoate 96%

Supplier: AMBEED, INC
Warning

A201498-10G A201498-1G A201498-250MG A201498-5G
76721-466EA 162 USD InStock
76721-466 76721-468 76721-470 76721-472
Methyl-3-methoxy-2-naphthoate 96%
Methyl 3-methoxy-2-naphthoate
Precaución: for Research Use Only


Formula: C₁₃H₁₂O₃
MW: 216.2300 g/mol
MDL Number: MFCD00454253
CAS Number: 13041-60-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty