Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-Boc-4-(2-bromo-ethyl)-piperidine 97%

Supplier: AMBEED, INC
Warning

A201879-100MG A201879-1G A201879-250MG A201879-5G
76721-482EA 19 USD InStock
76721-482 76721-484 76721-486 76721-488
N-Boc-4-(2-bromo-ethyl)-piperidine 97%
N-Boc-4-(2-bromo-ethyl)-piperidine
Precaución: for Research Use Only


Formula: C₁₂H₂₂BrNO₂
MW: 292.213 g/mol
MDL Number: MFCD06410598
CAS Number: 169457-73-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty