Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Piperidine-4-sulfonyl chloride, N-CBZ protected 95%

Supplier: AMBEED, INC
Danger

A203364-250MG A203364-10G
76721-510EA 23 USD InStock
76721-510 77667-700
Piperidine-4-sulfonyl chloride, N-CBZ protected 95%
Piperidine-4-sulfonyl chloride, N-CBZ protected
Precaución: for Research Use Only


Formula: C₁₃H₁₆ClNO₄S
MDL Number: MFCD05664659
CAS Number: 287953-54-2
UN: 3261
ADR: 8,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty