Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-phenylanthranilic acid 98%

Supplier: AMBEED, INC
Warning

A204397-100G A204397-10G A204397-25G A204397-5G
76721-516EA 31 USD InStock
76721-516 76721-518 77302-910 77667-710
N-phenylanthranilic acid 98%
N-phenylanthranilic acid
Precaución: for Research Use Only


Formula: C₁₃H₁₁NO₂
MW: 213.24 g/mol
Boiling Pt: 375
MDL Number: MFCD00002421
CAS Number: 91-40-7
Merck Index: 12,07427
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty