Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Pyrid-4-ylbenzoic acid 98%

Supplier: AMBEED, INC
Warning

A205431-1G A205431-250MG A205431-25G A205431-5G
76721-542EA 15 USD InStock
76721-542 76721-544 76721-546 76721-548
4-Pyrid-4-ylbenzoic acid 98%
4-Pyrid-4-ylbenzoic acid
Precaución: for Research Use Only


Formula: C₁₂H₉NO₂
MDL Number: MFCD04114574
CAS Number: 4385-76-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty