Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl methanesulphonate 98%

Supplier: AMBEED, INC
Danger

A206439-10G A206439-1G A206439-25G A206439-5G
76721-570EA 10 USD InStock
76721-570 76721-572 76721-574 76721-576
Methyl methanesulphonate 98%
Methyl methanesulphonate
Precaución: for Research Use Only


Formula: C₂H₆O₃S
MW: 110.13 g/mol
Boiling Pt: 203
Melting Pt: 20
Flash Pt: 104
MDL Number: MFCD00007557
CAS Number: 66-27-3
UN: 2810
ADR: 6.1 Ⅲ
Merck Index: 12,06170
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty