Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Phenanthrenol 95%

Supplier: AMBEED, INC
Warning

A211064-100MG A211064-1G A211064-250MG
76721-680EA 114 USD InStock
76721-680 76721-682 76721-684
3-Phenanthrenol 95%
3-Phenanthrenol
Precaución: for Research Use Only


Formula: C₁₄H₁₀O
MDL Number: MFCD00215969
CAS Number: 605-87-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty