Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(1H-Pyrazol-1-yl)methanol 97%

Supplier: AMBEED, INC
Warning

A211727-1G A211727-250MG A211727-5G
76721-722EA 40 USD InStock
76721-722 76721-724 76721-726
(1H-Pyrazol-1-yl)methanol 97%
(1H-Pyrazol-1-yl)methanol
Precaución: for Research Use Only


Formula: C₄H₆N₂O
MDL Number: MFCD00955584
CAS Number: 1120-82-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty