Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sodium isonicotinate 99%

Supplier: AMBEED, INC
Warning

A212554-25G A212554-5G A212554-100G
76721-752EA 20 USD InStock
76721-752 76721-754 77667-778
Sodium isonicotinate 99%
Sodium isonicotinate
Precaución: for Research Use Only


Formula: C₆H₄NNaO₂
MW: 145.0912 g/mol
MDL Number: MFCD00064868
CAS Number: 16887-79-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty