Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Fmoc-D-Cit-OH 97%

Supplier: AMBEED, INC
Warning

A213013-100MG A213013-250MG
76721-772EA 8 USD InStock
76721-772 76721-774
Fmoc-D-Cit-OH 97%
Fmoc-D-Cit-OH
Precaución: for Research Use Only


Formula: C₂₁H₂₃N₃O₅
MW: 397.431 g/mol
MDL Number: MFCD00151942
CAS Number: 200344-33-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty