Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-[Tris(hydroxymethyl)methyl]acrylamide 97%

Supplier: AMBEED, INC
Warning

A219111-10G A219111-1G A219111-25G A219111-5G
76721-908EA 129 USD InStock
76721-908 76721-910 76721-912 76721-914
N-[Tris(hydroxymethyl)methyl]acrylamide 97%
N-[Tris(hydroxymethyl)methyl]acrylamide
Precaución: for Research Use Only


Formula: C₇H₁₃NO₄
MDL Number: MFCD00081045
CAS Number: 13880-05-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty