Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methylsuccinic anhydride 98%

Supplier: AMBEED, INC
Warning

A220439-100G A220439-1G A220439-25G A220439-500G A220439-5G
76721-946EA 55 USD InStock
76721-946 76721-948 76721-950 76721-952 76721-954 76721-956
Methylsuccinic anhydride 98%
Methylsuccinic anhydride
Precaución: for Research Use Only


Formula: C₅H₆O₃
MDL Number: MFCD00005527
CAS Number: 4100-80-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty