Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Potassium thiophene-3-trifluoroborate 97%

Supplier: AMBEED, INC
Warning

A222039-1G A222039-25G A222039-5G
76721-992EA 15 USD InStock
76721-992 76721-994 76721-996
Potassium thiophene-3-trifluoroborate 97%
Potassium thiophene-3-trifluoroborate
Precaución: for Research Use Only


Formula: C₄H₃BF₃KS
MDL Number: MFCD03701626
CAS Number: 192863-37-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty