Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Potassium (2-nitrophenyl)trifluoroborate 96%

Supplier: AMBEED, INC
Warning

A222803-1G A222803-250MG A222803-5G
76722-010EA 22 USD InStock
76722-010 76722-012 76722-014
Potassium (2-nitrophenyl)trifluoroborate 96%
Potassium (2-nitrophenyl)trifluoroborate
Precaución: for Research Use Only


Formula: C₆H₄BF₃KNO₂
MDL Number: MFCD04115759
CAS Number: 850623-64-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty